Assay Details
Target Gene Details
Entrez Gene ID: | 52575 |
Gene Name: | tRNA methyltransferase 10C |
Gene Aliases: |
1300018J16Rik, D16Ertd454e, Rg9mtd1, Rnmtd1 |
Location: |
Chr.16:56033720-56037774 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trmt10c | NM_029092.3 | 2 | 795 | NP_083368.1 |
| AK005047.1 | 2 | 799 | BAB23773.1 | |
| AK087991.1 | 2 | 788 | BAC40080.1 | |
| AK135426.1 | 2 | 801 | BAE22529.1 | |
| AK166781.1 | 2 | 808 | BAE39015.1 | |
| BC023147.1 | 2 | 775 | AAH23147.1 | |
| BC106131.1 | 2 | 777 | AAI06132.1 |
More Information
Set Membership: |
|

Genomic Map