Assay Details
Target Gene Details
Entrez Gene ID: | 78913 |
Gene Name: | listerin E3 ubiquitin protein ligase 1 |
Gene Aliases: |
4930528H02Rik, AV266914, C87237, Listerin, Rnf160, Zfp294 |
Location: |
Chr.16:87376651-87432619 on Build GRCm38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ltn1 | NM_001081068.1 | 30 | 6964 | NP_001074537.1 |
| AK052360.1 | 1 | 1520 | ||
| BC027795.1 | 9 | 2934 | AAH27795.1 |
More Information
Set Membership: |
|

Genomic Map