Assay Details
Target Gene Details
Entrez Gene ID: | 76497 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 11 |
Gene Aliases: |
1500041B02Rik, AV117883, Hcgv, Tcte5, Tctex-5, Tctex5 |
Location: |
Chr.17:36948355-36951792 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r11 | NM_029632.3 | 1 | 306 | NP_083908.1 |
| AK005379.1 | 1 | 203 | BAB23986.1 | |
| AK077455.1 | 1 | 189 | ||
| AK089107.1 | 1 | 189 | BAC40754.1 | |
| AK162607.1 | 1 | 189 | ||
| AK170104.1 | 1 | 189 | BAE41566.1 | |
| BB842861.1 | 1 | 304 | ||
| BC027737.1 | 1 | 176 | AAH27737.1 | |
| EF528576.1 | ||||
| EF528577.1 | ||||
| EF528578.1 |
More Information
Set Membership: |
|

Genomic Map