Assay Details
Target Gene Details
Entrez Gene ID: | 76497 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 11 |
Gene Aliases: |
1500041B02Rik, AV117883, Hcgv, Tcte5, Tctex-5, Tctex5 |
Location: |
Chr.17:36948355-36951792 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r11 | NM_029632.3 | 3 | 1279 | NP_083908.1 |
| AK077455.1 | ||||
| AK162607.1 | 3 | 1163 |
More Information
Set Membership: |
|

Genomic Map