Assay Details
Target Gene Details
Entrez Gene ID: | 71302 |
Gene Name: | Rho GTPase activating protein 26 |
Gene Aliases: |
1810044B20Rik, 2610010G17Rik, 4933432P15Rik, AI853435, mKIAA0621 |
Location: |
Chr.18:38992968-39376285 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap26 | NM_175164.4 | 1 | 396 | NP_780373.3 |
| AK011368.1 | 1 | 399 | BAB27574.1 | |
| AK129176.1 | 1 | 334 | BAC97986.1 | |
| BC141394.1 | 1 | 265 |
More Information
Set Membership: |
|

Genomic Map