Assay Details
Target Gene Details
Entrez Gene ID: | 22195 |
Gene Name: | ubiquitin-conjugating enzyme E2L 3 |
Gene Aliases: |
C79827, UbcM4, Ubce7 |
Location: |
Chr.16:17152015-17201498 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ube2l3 | NM_009456.2 | 4 | 1857 | NP_033482.1 |
| AJ130961.1 | 4 | 1846 | CAA10265.1 | |
| AK165620.1 | 4 | 1838 | BAE38300.1 | |
| AK167847.1 | 4 | 1845 | BAE39867.1 | |
| AK208192.1 | ||||
| X97042.1 | 4 | 1857 | CAA65755.1 |
More Information
Set Membership: |
|

Genomic Map