Assay Details
Target Gene Details
Entrez Gene ID: | 225289 |
Gene Name: | expressed sequence AW554918 |
Gene Aliases: |
4921538P22, AI461742, Kiaa1328, mKIAA1328 |
Location: |
Chr.18:25169004-25467321 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AW554918 | NM_001033532.3 | 11 | 3502 | NP_001028704.2 |
| AK220161.1 | 5 | 2324 | BAD90347.1 | |
| BC066089.1 | 7 | 2969 | AAH66089.1 |

Genomic Map