Assay Details
Target Gene Details
Entrez Gene ID: | 224065 |
Gene Name: | urotensin 2B |
Gene Aliases: |
Gm538, U2B, Urp, Uts2d |
Location: |
Chr.16:27353322-27370239 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uts2b | NM_198166.3 | 5 | 698 | NP_937809.1 |
| AK131972.1 | 5 | 676 | BAE20912.1 | |
| AK131973.1 | 5 | 679 | BAE20913.1 | |
| AK132025.1 | 5 | 699 | BAE20950.1 | |
| AV144685.2 | 5 | 712 | ||
| AW490879.1 | 1 | 154 | ||
| AY321315.1 | 5 | 679 | AAQ83885.1 | |
| BB191600.1 | 1 | 194 | ||
| BB611261.1 | 5 | 699 |

Genomic Map