Assay Details
Target Gene Details
Entrez Gene ID: | 224997 |
Gene Name: | discs, large (Drosophila) homolog-associated protein 1 |
Gene Aliases: |
4933422O14Rik, 9630002F18, AI845682, AI848168, BB075781, D17Bwg0511e, GKAP/SAPAP, GKPA/SAPAP, Gkap, Sapap1, mKIAA4162 |
Location: |
Chr.17:69969078-70821413 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dlgap1 | NM_001128180.1 | NP_001121652.1 | ||
| NM_001128181.1 | 10 | 2780 | NP_001121653.1 | |
| NM_027712.3 | 10 | 3765 | NP_081988.3 | |
| NM_177639.6 | NP_808307.2 | |||
| AK016873.1 | ||||
| AK034182.1 | 5 | 1654 | BAC28619.1 | |
| AK147448.1 | BAE27919.1 | |||
| AK161941.1 | BAE36644.1 | |||
| AK220515.1 | BAD90519.1 | |||
| AY243846.2 | AAP70755.2 | |||
| BC062120.1 | AAH62120.1 | |||
| BC094369.1 | 9 | 2974 | AAH94369.1 |
More Information
Set Membership: |
|

Genomic Map