Assay Details
Target Gene Details
Entrez Gene ID: | 15043 |
Gene Name: | histocompatibility 2, T region locus 3 |
Gene Aliases: |
H-2T3, H2-Tw3, TL |
Location: |
Chr.17:36185571-36190151 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H2-T3 | NM_008208.4 | 6 | 1615 | NP_032234.3 |
| AK033602.1 | 6 | 1616 | BAC28383.1 | |
| BC125378.1 | ||||
| BC125380.1 | ||||
| X03051.1 | 5 | 1504 | CAA26859.1 |
More Information
Set Membership: |
|

Genomic Map