Assay Details
Target Gene Details
Entrez Gene ID: | 56462 |
Gene Name: | mitochondrial carrier homolog 1 (C. elegans) |
Gene Aliases: |
2310034O17Rik, AI255158, AU018396, C77849, PSAP |
Location: |
Chr.17:29332076-29347927 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtch1 | NM_019880.3 | 1 | 364 | NP_063933.1 |
| AF176007.2 | 1 | 364 | AAD52645.2 | |
| AF192558.2 | 1 | 364 | AAF12792.2 | |
| AK009623.1 | 1 | 247 | BAC25262.1 | |
| AK154010.1 | 1 | 220 | BAE32313.1 | |
| BC023135.1 | 1 | 207 | AAH23135.1 | |
| BC027274.1 | 1 | 213 | AAH27274.1 |
More Information
Set Membership: |
|

Genomic Map