Assay Details
Target Gene Details
Entrez Gene ID: | 56468 |
Gene Name: | suppressor of cytokine signaling 5 |
Gene Aliases: |
1810018L08Rik, Cish5, mKIAA0671 |
Location: |
Chr.17:87107407-87137647 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Socs5 | NM_019654.2 | 3 | 3066 | NP_062628.2 |
| AK007534.1 | 1 | 578 | ||
| AK172997.1 | 1 | 1610 | BAD32275.1 | |
| BC053015.1 | 3 | 3066 | AAH53015.1 |
More Information
Set Membership: |
|

Genomic Map