Assay Details
Target Gene Details
Entrez Gene ID: | 381126 |
Gene Name: | GRB2 associated, regulator of MAPK1 |
Gene Aliases: |
C86169, Fam59a, Gm944, mKIAA4238 |
Location: |
Chr.18:21127342-21300139 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Garem | NM_001033445.2 | 1 | 88 | NP_001028617.2 |
| AK148318.1 | 1 | 89 | BAE28477.1 | |
| AK220564.1 | 1 | 72 | BAD90542.1 |

Genomic Map