Assay Details
Target Gene Details
Entrez Gene ID: | 320213 |
Gene Name: | SUMO/sentrin specific peptidase 5 |
Gene Aliases: |
6230429P13Rik, A730063F07Rik, AI851888, BB189556, SMT3IP3 |
Location: |
Chr.16:31959670-32010740 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Senp5 | NM_177103.4 | 1 | 247 | NP_796077.2 |
| AK028177.1 | 1 | 248 | ||
| AK042087.1 | 1 | 139 | ||
| AK043171.1 | 1 | 132 | BAC31483.1 | |
| AK158338.1 | 1 | 112 | ||
| AY522918.1 | 1 | 111 | AAT01901.1 | |
| BC067014.1 | AAH67014.1 | |||
| BC080830.1 | 1 | 135 | AAH80830.1 |
More Information
Set Membership: |
|

Genomic Map