Assay Details
Target Gene Details
Entrez Gene ID: | 320213 |
Gene Name: | SUMO/sentrin specific peptidase 5 |
Gene Aliases: |
6230429P13Rik, A730063F07Rik, AI851888, BB189556, SMT3IP3 |
Location: |
Chr.16:31959670-32010740 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Senp5 | NM_177103.4 | 10 | 5687 | NP_796077.2 |
| AK018100.1 | ||||
| AK078133.1 | 1 | 1568 |
More Information
Set Membership: |
|

Genomic Map