Assay Details
Target Gene Details
Entrez Gene ID: | 271377 |
Gene Name: | zinc finger and BTB domain containing 11 |
Gene Aliases: |
9230110G02Rik, ZNF-U69274 |
Location: |
Chr.16:55972697-56008913 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zbtb11 | NM_173026.2 | 1 | 291 | NP_766614.2 |
Target Gene Details
Entrez Gene ID: | 66391 |
Gene Name: | zinc finger and BTB domain containing 11, opposite strand 1 |
Gene Aliases: |
2310061J03Rik |
Location: |
Chr.16:55973268-55974617 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zbtb11os1 | NR_027965.1 | 1 | 524 | |
| AK006543.1 | 1 | 323 | ||
| AK010004.1 | 1 | 501 | ||
| BC017620.1 | 1 | 457 | AAH17620.1 |
More Information
Set Membership: |
|

Genomic Map