Assay Details
Target Gene Details
Entrez Gene ID: | 73242 |
Gene Name: | alpha tubulin acetyltransferase 1 |
Gene Aliases: |
0610011P08Rik, 2610008K08Rik, 2610110G12Rik, 3110080J08Rik, Mec17, TAT |
Location: |
Chr.17:35897595-35910093 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atat1 | NM_001142744.1 | 13 | 1349 | NP_001136216.1 |
AK011350.1 | 11 | 1211 | ||
AK011750.2 | 13 | 1325 | ||
AK011838.1 | 13 | 1340 | ||
AK028297.1 | 12 | 1310 | BAC25866.1 | |
BC028847.1 | 13 | 1328 | AAH28847.1 | |
BC030669.1 | 12 | 1258 | AAH30669.1 | |
BQ748472.1 | 4 | 385 |
