Assay Details
Target Gene Details
Entrez Gene ID: | 19274 |
Gene Name: | protein tyrosine phosphatase, receptor type, M |
Gene Aliases: |
RPTPmu, mKIAA4044 |
Location: |
Chr.17:66666848-67354541 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptprm | NM_008984.2 | 12 | 2519 | NP_033010.2 |
| AK039261.1 | 3 | 652 | ||
| AK129005.1 | 10 | 1886 | ||
| AK220171.1 | 10 | 1790 | BAD90356.1 | |
| BC058614.1 | 12 | 2529 | AAH58614.1 | |
| BC079621.1 | 12 | 2133 | AAH79621.1 | |
| X58287.1 | 12 | 2444 | CAA41225.1 |
More Information
Set Membership: |
|

Genomic Map