Assay Details
Target Gene Details
Entrez Gene ID: | 100503704 |
Gene Name: | predicted gene, 16861 |
Gene Aliases: |
- |
Location: |
Chr.16:4935164-4939585 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm16861 | NR_037666.1 | 1 | 123 | |
| AK078615.2 | 1 | 125 |
Target Gene Details
Entrez Gene ID: | 66911 |
Gene Name: | nudix (nucleoside diphosphate linked moiety X)-type motif 16-like 1 |
Gene Aliases: |
1110001K21Rik, 5330437I08Rik, Sdos |
Location: |
Chr.16:4939111-4941021 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nudt16l1 | NM_025839.3 | 2 | 269 | NP_080115.1 |
| AF435792.1 | 2 | 281 | AAL57490.1 | |
| AK002378.1 | 2 | 262 | BAB22055.1 | |
| AK003241.1 | 2 | 270 | BAB22662.1 | |
| AK007897.1 | 2 | 265 | BAB25336.1 | |
| AK019936.1 | 2 | 272 | ||
| AK159841.1 | 2 | 262 | BAE35419.1 | |
| AK186853.1 | ||||
| AK188290.1 | ||||
| BC025569.1 | 2 | 268 | AAH25569.1 | |
| BC028494.1 | 2 | 262 | AAH28494.1 |

Genomic Map