Assay Details
Target Gene Details
Entrez Gene ID: | 66911 |
Gene Name: | nudix (nucleoside diphosphate linked moiety X)-type motif 16-like 1 |
Gene Aliases: |
1110001K21Rik, 5330437I08Rik, Sdos |
Location: |
Chr.16:4939111-4941021 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nudt16l1 | NM_025839.3 | 3 | 583 | NP_080115.1 |
| AF435792.1 | 3 | 595 | AAL57490.1 | |
| AK002378.1 | 3 | 576 | BAB22055.1 | |
| AK003241.1 | 3 | 584 | BAB22662.1 | |
| AK007897.1 | 3 | 579 | BAB25336.1 | |
| AK019936.1 | ||||
| AK159841.1 | 3 | 576 | BAE35419.1 | |
| AK186853.1 | 2 | 293 | ||
| AK206496.1 | 1 | 89 | ||
| BC025569.1 | 3 | 657 | AAH25569.1 | |
| BC028494.1 | AAH28494.1 |

Genomic Map