Assay Details
Target Gene Details
Entrez Gene ID: | 50498 |
Gene Name: | Epstein-Barr virus induced gene 3 |
Gene Aliases: |
EBI-3, IL-27 |
Location: |
Chr.17:55952623-55957022 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ebi3 | NM_015766.2 | 1 | 199 | NP_056581.1 |
| AF013114.1 | 1 | 182 | AAB67115.1 | |
| AK155023.1 | 1 | 162 | BAE32996.1 | |
| AK155903.1 | 1 | 54 | BAE33494.1 | |
| BC008209.1 | 1 | 147 | AAH08209.1 | |
| BY134465.1 | 1 | 200 | ||
| CX240802.1 | 1 | 146 |
Target Gene Details
Entrez Gene ID: | 240120 |
Gene Name: | zinc finger protein 119b |
Gene Aliases: |
- |
Location: |
Chr.17:55938373-55956178 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp119b |
More Information
Set Membership: |
|

Genomic Map