Assay Details
Target Gene Details
Entrez Gene ID: | 70536 |
Gene Name: | glutaminyl-peptide cyclotransferase (glutaminyl cyclase) |
Gene Aliases: |
5730422A13Rik |
Location: |
Chr.17:79051906-79090268 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Qpct | NM_027455.2 | 1 | 239 | NP_081731.1 |
| AK045974.1 | 1 | 240 | BAC32556.1 | |
| BC151027.1 | ||||
| BC151029.1 |
More Information
Set Membership: |
|

Genomic Map