Assay Details
Target Gene Details
Entrez Gene ID: | 74196 |
Gene Name: | tetratricopeptide repeat domain 27 |
Gene Aliases: |
2610511O17Rik |
Location: |
Chr.17:74717736-74863570 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc27 | NM_152817.4 | 1 | 165 | NP_690030.3 |
| AK012131.1 | 1 | 165 | ||
| AK030942.1 | 1 | 166 | BAC27189.1 | |
| AK049324.1 | 1 | 166 | BAC33683.1 | |
| AK168138.1 | 1 | 156 | BAE40106.1 |

Genomic Map