Assay Details
Target Gene Details
Entrez Gene ID: | 12475 |
Gene Name: | CD14 antigen |
Gene Aliases: |
- |
Location: |
Chr.18:36725064-36726815 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd14 | NM_009841.4 | 2 | 777 | NP_033971.1 |
| AK149744.1 | 2 | 704 | BAE29057.1 | |
| AK151139.1 | 1 | 75 | BAE30146.1 | |
| AK152427.1 | 2 | 566 | ||
| AK153229.1 | 2 | 567 | BAE31822.1 | |
| AK156262.1 | 2 | 566 | BAE33646.1 | |
| BC057889.1 | 2 | 616 | AAH57889.1 | |
| CT010355.1 | 1 | 453 | CAJ18563.1 | |
| X13333.1 | 2 | 485 | CAA31710.1 |
More Information
Set Membership: |
|

Genomic Map