Assay Details
Target Gene Details
Entrez Gene ID: | 72020 |
Gene Name: | zinc finger protein 654 |
Gene Aliases: |
1600021C16Rik, 1810008K20Rik, Znf654 |
Location: |
Chr.16:64780347-64851905 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp654 | NM_028059.2 | 2 | 4256 | NP_082335.1 |
| AK007382.1 | 1 | 783 | ||
| BC027760.1 | 2 | 3958 | AAH27760.1 | |
| BC068018.1 | 2 | 4256 | AAH68018.1 |
More Information
Set Membership: |
|

Genomic Map