Assay Details
Target Gene Details
Entrez Gene ID: | 225049 |
Gene Name: | tetratricopeptide repeat domain 7 |
Gene Aliases: |
1110035E02Rik, 1700007L07Rik, Ttc7a, fsn, hea |
Location: |
Chr.17:87282082-87381770 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc7 | NM_028639.3 | 20 | 3644 | NP_082915.1 |
| AK040578.1 | 20 | 3647 | BAC30634.1 | |
| AK040610.1 | 20 | 3615 | BAC30643.1 | |
| AK050263.1 | 20 | 3631 | BAC34153.1 | |
| AK080284.1 | 20 | 3623 | BAC37865.1 | |
| AK165244.1 | 13 | 2306 | BAE38100.1 | |
| BC042512.1 | 20 | 3321 | AAH42512.2 | |
| BC049254.1 | 4 | 1367 | AAH49254.1 |

Genomic Map