Assay Details
Target Gene Details
Entrez Gene ID: | 224022 |
Gene Name: | solute carrier family 7 (cationic amino acid transporter, y+ system), member 4 |
Gene Aliases: |
AI853530 |
Location: |
Chr.16:17572018-17576892 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc7a4 | NM_144852.3 | 3 | 1261 | NP_659101.2 |
| AK030586.1 | 3 | 1265 | BAC27031.1 | |
| AK043772.1 | 3 | 1262 | BAC31651.1 | |
| AK162398.1 | 3 | 1305 | BAE36893.1 | |
| BC016100.1 | AAH16100.1 |
More Information
Set Membership: |
|

Genomic Map