Assay Details
Target Gene Details
Entrez Gene ID: | 12703 |
Gene Name: | suppressor of cytokine signaling 1 |
Gene Aliases: |
Cish1, Cish7, JAB, SOCS-1, SSI-1 |
Location: |
Chr.16:10783808-10785536 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Socs1 | NM_001271603.1 | 2 | 599 | NP_001258532.1 |
| NM_009896.2 | 2 | 479 | NP_034026.1 | |
| AB000677.1 | 1 | 323 | BAA21538.1 | |
| AB000710.1 | 1 | 377 | BAA21539.1 | |
| AF120490.1 | 2 | 452 | AAD24777.1 | |
| AF180302.1 | 2 | 457 | AAD53324.1 | |
| BC132366.1 | 2 | 476 | ||
| BC132368.1 | 2 | 476 | ||
| U88325.1 | 2 | 441 | AAB62400.1 |
More Information
Set Membership: |
|

Genomic Map