Assay Details
Target Gene Details
Entrez Gene ID: | 78923 |
Gene Name: | chondroitin sulfate synthase 3 |
Gene Aliases: |
4833446K15Rik, AI662215 |
Location: |
Chr.18:59175340-59411336 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chsy3 | NM_001081328.1 | 1 | 385 | NP_001074797.1 |
| AK038429.1 | 1 | 325 | ||
| AK043732.1 | 1 | 384 | BAC31633.1 | |
| AK044099.1 | 1 | 384 | BAC31776.1 | |
| AK048196.1 | 1 | 296 | ||
| BC151164.1 | 1 | 75 | ||
| BC151165.1 | 1 | 75 | ||
| BC157985.1 | 1 | 75 |
More Information
Set Membership: |
|

Genomic Map