Assay Details
Target Gene Details
Entrez Gene ID: | 78923 |
Gene Name: | chondroitin sulfate synthase 3 |
Gene Aliases: |
4833446K15Rik, AI662215 |
Location: |
Chr.18:59175340-59411336 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chsy3 | NM_001081328.1 | 2 | 1384 | NP_001074797.1 |
| AK019528.1 | 2 | 371 | BAC25596.1 | |
| AK038429.1 | 2 | 1325 | ||
| AK048196.1 | 2 | 1296 | ||
| BC151164.1 | 2 | 1074 | ||
| BC151165.1 | 2 | 1074 | ||
| BC157985.1 | 2 | 1074 |
More Information
Set Membership: |
|

Genomic Map