Assay Details
Target Gene Details
Entrez Gene ID: | 17075 |
Gene Name: | epithelial cell adhesion molecule |
Gene Aliases: |
CD326, EGP, EGP-2, Egp314, Ep-CAM, EpCAM1, GA733-2, Ly74, TROP1, Tacsd1, Tacstd1, gp40 |
Location: |
Chr.17:87635979-87651127 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epcam | NM_008532.2 | 2 | 162 | NP_032558.2 |
| BI986346.1 | 2 | 162 |

Genomic Map