Assay Details
Target Gene Details
Entrez Gene ID: | 240028 |
Gene Name: | leucyl/cystinyl aminopeptidase |
Gene Aliases: |
2010309L07Rik, 4732490P18Rik, CAP, IRAP, PLAP, gp160, vp165 |
Location: |
Chr.17:17527723-17624489 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lnpep | NM_172827.3 | 2 | 234 | NP_766415.1 |
| AK029094.1 | 2 | 235 | BAC26293.1 | |
| AK036967.1 | 2 | 235 | BAC29651.1 | |
| BC066791.1 | 3 | 558 | AAH66791.1 | |
| BC120925.1 | 2 | 191 | ||
| BC120926.1 | 2 | 191 |
More Information
Set Membership: |
|

Genomic Map