Assay Details
Target Gene Details
Entrez Gene ID: | 19725 |
Gene Name: | regulatory factor X, 2 (influences HLA class II expression) |
Gene Aliases: |
5430432H19Rik |
Location: |
Chr.17:56775897-56831054 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rfx2 | NM_009056.2 | 17 | 2500 | NP_033082.1 |
| NM_027787.1 | 18 | 2575 | NP_082063.1 | |
| AK044439.1 | 18 | 2575 | BAC31919.1 | |
| BC004654.1 | 17 | 2517 | AAH04654.1 | |
| X76089.1 | 17 | 2455 | CAA53703.1 |
More Information
Set Membership: |
|

Genomic Map