Assay Details
Target Gene Details
Entrez Gene ID: | 171233 |
Gene Name: | vomeronasal 1 receptor 235 |
Gene Aliases: |
V1rf2 |
Location: |
Chr.17:21260427-21262863 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vmn1r235 | NM_134199.3 | 1 | 94 | NP_598960.3 |
| AF394953.1 | 1 | 94 | AAM69673.1 |
More Information
Set Membership: |
|

Genomic Map