Assay Details
Target Gene Details
Entrez Gene ID: | 171234 |
Gene Name: | vomeronasal 1 receptor 237 |
Gene Aliases: |
V1rf3, Vfrf3 |
Location: |
Chr.17:21314017-21314886 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vmn1r237 | NM_134200.1 | 1 | 576 | NP_598961.1 |
| BC125543.1 | 1 | 630 | ||
| BC138066.1 | 1 | 630 |
More Information
Set Membership: |
|

Genomic Map