Assay Details
Target Gene Details
Entrez Gene ID: | 106878 |
Gene Name: | small integral membrane protein 3 |
Gene Aliases: |
2010002N04Rik, AI195350, AW561905, Nid67, cI-41 |
Location: |
Chr.18:60474191-60518947 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smim3 | NM_134133.2 | 2 | 1287 | NP_598894.1 |
| AF313412.1 | 2 | 829 | AAG53958.1 | |
| AK143383.1 | 2 | 1291 | BAE25359.1 | |
| AK155873.1 | 2 | 933 | BAE33475.1 | |
| AK168283.1 | 2 | 1111 | BAE40229.1 | |
| AK171810.1 | 2 | 945 | BAE42676.1 | |
| BC012898.1 | 2 | 795 | AAH12898.1 |

Genomic Map