Assay Details
Target Gene Details
Entrez Gene ID: | 14962 |
Gene Name: | complement factor B |
Gene Aliases: |
AI195813, AI255840, B, Bf, C2, Fb, H2-Bf |
Location: |
Chr.17:34856374-34862514 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cfb | NM_001142706.1 | 1 | 74 | NP_001136178.1 |
| NM_008198.2 | 1 | 74 | NP_032224.2 | |
| CA492475.1 | 1 | 111 |

Genomic Map