Assay Details
Target Gene Details
Entrez Gene ID: | 225600 |
Gene Name: | phosphodiesterase 6A, cGMP-specific, rod, alpha |
Gene Aliases: |
Pdea, nmf282 |
Location: |
Chr.18:61220080-61289750 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pde6a | NM_146086.2 | 22 | 3423 | NP_666198.1 |
| AK042142.1 | 6 | 1283 | BAC31183.1 | |
| BC044892.1 | 22 | 3423 | AAH44892.1 |
More Information
Set Membership: |
|

Genomic Map