Assay Details
Target Gene Details
Entrez Gene ID: | 11516 |
Gene Name: | adenylate cyclase activating polypeptide 1 |
Gene Aliases: |
PACAP |
Location: |
Chr.17:93198904-93206541 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adcyap1 | NM_009625.2 | 5 | 1837 | NP_033755.1 |
| AK079530.1 | 5 | 1815 | BAC37673.1 | |
| AK142765.1 | 3 | 1683 | ||
| BC057344.1 | 5 | 1837 | AAH57344.1 | |
| D14716.1 | 4 | 1636 |
More Information
Set Membership: |
|

Genomic Map