Assay Details
Target Gene Details
Entrez Gene ID: | 225020 |
Gene Name: | fasciculation and elongation protein zeta 2 (zygin II) |
Gene Aliases: |
9030616F16, D17Ertd315e |
Location: |
Chr.17:78369212-78418152 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fez2 | NM_001285940.1 | 5 | 914 | NP_001272869.1 |
| NM_001285946.1 | 5 | 914 | NP_001272875.1 | |
| NM_001285949.1 | 5 | 914 | NP_001272878.1 | |
| NM_199448.3 | 5 | 914 | NP_955519.1 | |
| AK033547.1 | 5 | 856 | ||
| AK150241.1 | 5 | 880 | BAE29405.1 | |
| AK157221.1 | 5 | 881 | BAE34006.1 | |
| AK159545.1 | 5 | 867 | BAE35171.1 | |
| AK165595.1 | 5 | 864 | BAE38283.1 | |
| AK177083.1 | ||||
| AY382580.1 | 5 | 865 | AAR36860.1 | |
| BC064130.1 | 5 | 820 | AAH64130.1 | |
| BC096619.1 | 5 | 859 | AAH96619.1 |

Genomic Map