Assay Details
Target Gene Details
Entrez Gene ID: | 56299 |
Gene Name: | FK506 binding protein-like |
Gene Aliases: |
DIR1, NG7, Ppiase-X, Wisp39 |
Location: |
Chr.17:34639574-34646327 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fkbpl | NM_019873.2 | 2 | 790 | NP_063926.1 |
| AK139541.1 | 1 | 370 | ||
| AK146018.1 | 2 | 860 | BAE26836.1 | |
| BC046338.1 | 2 | 790 | AAH46338.1 |
More Information
Set Membership: |
|

Genomic Map