Assay Details
Target Gene Details
Entrez Gene ID: | 15200 |
Gene Name: | heparin-binding EGF-like growth factor |
Gene Aliases: |
AW047313, Dtr, Dts, Hegfl |
Location: |
Chr.18:36504927-36515805 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hbegf | NM_010415.2 | 6 | 2138 | NP_034545.1 |
| AK145548.1 | 6 | 2107 | BAE26501.1 | |
| AW489543.1 | 1 | 255 |
More Information
Set Membership: |
|

Genomic Map