Assay Details
Target Gene Details
Entrez Gene ID: | 20229 |
Gene Name: | spermidine/spermine N1-acetyl transferase 1 |
Gene Aliases: |
AA617398, SSAT, Sat |
Location: |
Chr.X:155213126-155216449 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sat1 | NM_001291865.1 | 1 | 92 | NP_001278794.1 |
| NM_009121.4 | 1 | 92 | NP_033147.1 | |
| AK002531.1 | BAB22167.1 | |||
| AK154445.1 | BAE32591.1 | |||
| BC058696.1 | 1 | 52 | AAH58696.1 | |
| BU699066.1 | 1 | 92 |
More Information
Set Membership: |
|

Genomic Map