Assay Details
Target Gene Details
Entrez Gene ID: | 53412 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 3C |
Gene Aliases: |
PTG, Ppp1r5 |
Location: |
Chr.19:36731731-36736604 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r3c | NM_016854.2 | 2 | 499 | NP_058550.1 |
| AK078506.1 | 1 | 86 | BAC37313.1 | |
| AK078735.1 | 2 | 544 | ||
| BC052769.1 | 2 | 548 | AAH52769.1 | |
| BC054739.1 | 2 | 499 | AAH54739.1 | |
| U89924.2 | 1 | 452 | AAB49689.1 |
More Information
Set Membership: |
|

Genomic Map