Assay Details
Target Gene Details
Entrez Gene ID: | 12361 |
Gene Name: | calcium/calmodulin-dependent serine protein kinase (MAGUK family) |
Gene Aliases: |
DXPri1, DXRib1, LIN-2, Pals3, mLin-2 |
Location: |
Chr.X:13517080-13846784 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cask | NM_001284503.1 | 27 | 4849 | NP_001271432.1 |
| NM_001284504.1 | 24 | 4711 | NP_001271433.1 | |
| NM_001284505.1 | 26 | 4831 | NP_001271434.1 | |
| NM_009806.3 | 25 | 4762 | NP_033936.2 |
More Information
Set Membership: |
|

Genomic Map