Assay Details
Target Gene Details
Entrez Gene ID: | 108673 |
Gene Name: | coiled-coil domain containing 86 |
Gene Aliases: |
4933411H20Rik, 6720480F16Rik, D19Ertd678e |
Location: |
Chr.19:10941481-10949266 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccdc86 | NM_023731.3 | 4 | 1804 | NP_076220.2 |
AK036812.1 | 4 | 1795 |
