Assay Details
Target Gene Details
Entrez Gene ID: | 171486 |
Gene Name: | CD99 antigen-like 2 |
Gene Aliases: |
AW548191, Mic2l1, Xap89 |
Location: |
Chr.X:71420060-71492869 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd99l2 | NM_001199349.1 | 10 | 2251 | NP_001186278.1 |
| NM_138309.3 | 9 | 2182 | NP_612182.1 | |
| AK009181.1 | 1 | 385 | ||
| AK031264.1 | 10 | 2008 | ||
| AK080225.1 | 10 | 2007 | BAC37853.1 | |
| AK166275.1 | 10 | 2002 | BAE38674.1 | |
| AK209296.1 | ||||
| AK211589.1 | ||||
| AK213435.1 | ||||
| AK215094.1 | ||||
| AK215804.1 | ||||
| AK219848.1 | ||||
| AY078163.1 | 9 | 1978 | AAL86615.1 |

Genomic Map