Assay Details
Target Gene Details
Entrez Gene ID: | 245347 |
Gene Name: | predicted pseudogene 4984 |
Gene Aliases: |
AI428901, EG245347 |
Location: |
Chr.X:12651883-12652821 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm4984 | NM_001101484.1 | 1 | 162 | NP_001094954.1 |

Genomic Map