Assay Details
Target Gene Details
Entrez Gene ID: | 194744 |
Gene Name: | solute carrier family 25, member 43 |
Gene Aliases: |
Gm8 |
Location: |
Chr.X:36743549-36777307 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a43 | NM_001085497.2 | 1 | 263 | NP_001078966.1 |
| NR_075096.1 | 1 | 263 | ||
| NR_075097.1 | 1 | 263 | ||
| AK153028.1 | 1 | 186 | ||
| BC147709.1 | 1 | 263 | ||
| BC147717.1 | 1 | 263 |
More Information
Set Membership: |
|

Genomic Map