Assay Details
Target Gene Details
Entrez Gene ID: | 75514 |
Gene Name: | RIKEN cDNA 1700013H16 gene |
Gene Aliases: |
Slx2, Xlr6 |
Location: |
Chr.X:53742901-53757970 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700013H16Rik | NM_001200013.1 | 9 | 992 | NP_001186942.1 |
| AK005953.1 | 9 | 975 | BAB24336.1 |

Genomic Map